Wearing lab safety equipment is extremely important because it protects researchers and lab employees from injuries in the workplace. Depending on the research or lab studies being conducted, there could be a chance of exposure to hazardous materials. By wearing proper attire and safety equipment, the risk of harm can be minimized, and labs can help prevent accidents from occurring.
Safety
Nothing that happens in the workplace is as important as your health and safety. Avantor® helps keep you and your colleagues safe with a complete line of safety products and personal protective equipment.
Toutes les fournitures de sécurité pour laboratoire dont vous avez besoin, y compris les absorbants, les conteneurs d'élimination, les systèmes d'étiquetage et plus encore.
Une gamme complète d'équipements de protection individuelle (EPI) de la tête aux pieds, fabriqués par les leaders de l'industrie.
Une gamme complète de produits de manutention et de stockage des produits chimiques, d'entretien des installations et de gestion des déchets provenant de marques auxquelles vous faites confiance.
Avantor Safety and PPE Products
Nothing that happens in the workplace is as important as your health and safety. Avantor® helps keep you and your colleagues safe with a complete line of safety products and personal protective equipment.
Featured Supplier Brands
Browse safety supplies and personal protective equipment from industry manufacturers. From the lab to the production floor, warehouse, cleanroom and everywhere at your site, Avantor has the PPE, cleaning and safety solutions you need to keep your facilities running safely and without interruption.
What Is the Importance of Wearing Lab Safety Equipment?
What Is the Essential Safety Equipment Needed in a Lab?
Some of the typical lab safety equipment needed in most labs includes:
- Chemical fume hoods – Fume hoods draw the harmful vapors out of the laboratory to help prevent you from inhaling vaporous chemicals and gases.
- Eyewash stations – A nearby eyewash station can make a big difference if any substance gets into your eyes from a splash or other accident.
- Fire blankets – Fire blankets can be used to smother the flames if someone’s clothes catch fire.
- Fire extinguishers – Fire can develop fires when labs use flammable materials or electrical equipment. Labs must have easily accessible fire extinguishers to extinguish flames immediately.
- First aid kits – Multiple well-stocked first aid kits should be available in every lab for use in the event of an injury.
- Laboratory coats – Covering your body with a lab coat can help prevent hazardous materials from contacting your skin.
- Laboratory-safe refrigerators – Lab-safe fridges are specifically designed to have no ignition sources. They also have other safety features that ensure the safe storage of lab materials.
- Protective gloves – Lab work requires working with your hands, and protective gloves offer a reduced risk of injury.
- Safety goggles – Proper lab eyewear helps protect your eye.
- Safety showers – A safety shower lets you quickly wash off harmful substances if they splash onto your skin.
Avantor Lets Your Lab Focus on Innovation.
Safety is a top priority in your workplace. From safety equipment to safety information, you can depend on Avantor to provide it all.
Resources to Improve Safety
Découvrez les produits de sécurité, les dernières tendances et des conseils pratiques pour renforcer la sécurité au sein de votre laboratoire.
Les experts d'Ansell GUARDIAN, un service de conseil, aident les entreprises à sélectionner et à mettre en œuvre les solutions d'EPI appropriées pour améliorer la sécurité, accroître la productivité et réduire les coûts.
Les experts d'Ansell GUARDIAN sont des experts en EPI.