Order Entry
daily-lab-consumables-955x590.png
Products for everyday use
The Avantor® collection of products gives you an easy choice of essential labware. You can easily see a wide selection of quality products to keep your lab running efficiently and effectively.
Our consumables, including beakers, test tubes, cuvettes, lab notebooks, glassware and more, are ideal for a variety of applications. These critical supplies deliver quality, value and convenience so you can focus on what matters most: your work.
8611077-bottles-540x390.jpg
Bottles made of HDPE, PP, PE, glass amber, and more. Narrow or wide mouth with tamper-evident seals and screws, snaps or dropper caps/closures.
AT-Plain-Round-Carboy-Group-540x390.jpg
Ideal for storing and dispensing water, media and other solutions in the lab.
21008-103-centrifuge-tubes-540x390.jpg
PS, PP or PTFE tubes, non-sterile or sterile. DNase-, RNase, pyrogen- and TSE-/BSE-free options are available in racks and bags.
T_2015_-198_Int-thread-cryo-vials-540x390.jpg
Vials for storage of biological material, including human or animal cells, at temperatures as low as -197 °C.
516-0861-filtration-540x390.jpg
Filter papers, syringe filters, bottle-top filters and more for chemistry, chromatography and life science filtration.
631-0113-microscope-slides-540x390.jpg
Slides and coverslips for basic biology education, health care diagnostics and more.
VWR-Screw-Cap-MCT-540x390.jpg
Snap-cap and screw-cap tubes from 0.5 - 2.0 ml in a variety of colors.
VWRI391-0556-petri-dishes-540x390.jpg
Gamma-irradiated, sterile or aseptic PS dishes in a variety of sizes and formats.
VWRI613-1052-pipette-tips-540x390.jpg
Designed to fit volumes ranging from 0.1 - 1250 µl and efficiently draw and deliver required liquids for any application with high accuracy and precision.
Serological-Pipettes-540x390.jpg
Plastic or glass transfer/Pasteur and serological pipettes in a variety of volumes, formats and quantities.
613-1174-reservoirs-540x390.jpg
These containers feature a trough design, graduation marks on interior walls and pour-off spouts on each corner to simplify operations.
VWRI212-0773-test-tubes-540x390.jpg
These high-quality glass and plastic test tubes are designed and constructed for optimal performance and excellent resistance to industrial scientific chemicals.
612.6709-vol-flask-540x390.jpg
Select models of borosilicate glass, Nalgene or other plastics that conform to Class A or Class B requirements.
611-1374-weighing-dishes-540x390.jpg
Paper, plastic, aluminum or glass, these weighing boats, scoops and dishes are available in many volumes down to the micro range. Ideal for weighing and moisture determination.
Contact us today to discuss your needs. Our team is ready to help.