Upstream processing for cell therapy
Compared to production volumes associated with other biologics (e.g., mAbs: 2,000L to 10,000L), cell therapy production volumes are more commonly between 1L to 10L. Even the quality parameters are different.
Instead of striving for maximum titers and protein quality, cell therapies focus on maximum cell dose for delivery and the functionality of the cells/tissues being administered.
Thus, upstream production parameters covering media, supplements, bioreactor systems, and starting cell quality are very important. In the end, when the culture process has been completed, therapeutic cells need to be harvested employing cell dissociation reagents that do not impact the quality of the cell product.
Explore solutions for upstream processing of cell therapy
Product choices for discovery, product development & pre-clinical
Product choices for scale-up & production
Ready to Get Started?
Ready to Maximize Production Efficiency?
Our Bioprocessing Solutions team can help you streamline your processes.