eVol® XR
Enabled analytical syringe
The world’s first analytical syringe
- XCHANGE® syringes are easily and quickly switched allowing them to be dedicated to individual liquids or methods to prevent possible cross-contamination of reagents
- eVol® is ergonomic, comfortable and easy to use
- eVol® is easily calibrated and calibration factors are saved for each syringe, enabling laboratories to comply with stringent global laboratory standards (e.g. GLP, GMP, FDA)
- eVol® is suitable for use with volatile samples
- eVol® is programmable to store a laboratory workflow (up to 98 steps)
- eVol® is suitable for direct injection onto a chromatography column with a consistent flow rate