Welcome to your Agilent Newsletter
In this edition of our Agilent Newsletter we look at some of the products in the Agilent AdvanceBio range of HPLC columns.
Agilent AdvanceBio columns are designed and produced to provide results you can be sure of when analyzing highly complex biotherapeutic molecules and monitoring their purity, potency, and other critical quality attributes. Contact us for more information on this range and see how we can help with your application
Agilent AdvanceBio Glycan Mapping
Agilent AdvanceBio Glycan Mapping Columns are designed and manufactured to deliver fast, high resolution, reproducible glycan identification using HILIC chromatography. AdvanceBio Glycan Mapping columns leverage technology that optimises results for MS and fluorescence detection. Choose from two UHPLC : 2.7 µm superficially porous, for high resolution configurationsand lower backpressure, or 1.8 µm for highest resolution.
    Request Quote
    Agilent AdvanceBio Amino Acid Analysis (AAA)
    Agilent’s AdvanceBio Amino Acid Analysis (AAA) delivers fast, sensitive, and reproducible separations of amino acids in protein hydrolysates and cell culture media.
    AdvanceBio AAA includes proven reagents for amino acid derivatisation, a ready-to-use amino acid standards kit and columns based on Agilent’s innovative Poroshell technology.
      Request Quote
      Additional Information
      Agilent AdvanceBio RP-mAb
      Agilent AdvanceBio RP-mAb columns are designed to optimise the performance of intact and reduced mAb analysis when analysing monoclonal antibodies for biopharma discovery, development, and QA/QC applications.
      Analysis of intact and reduced monoclonal antibodies are critical measurements for characterising therapeutic proteins and understanding their efficacy and stability
        Request Quote
        Additional Information
        Agilent AdvanceBio SEC
        AdvanceBio SEC columns deliver accurate, precise quantitation for mAb aggregation and protein analysis. These innovative size exclusion chromatography columns were designed and engineered by Agilent to improve lab productivity by providing robust, reliable methods that eliminate sample reanalysis.
          Request Quote
          Additional Information