GF Signet Piping System Metering Ball Valve, Polypropylene, 20 mm Socket
MFLX01360-45
Precise control over 180° range.
- Choose between PVC or polypropylene (PP) construction
Ball valves give you precise control over the entire 180° range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5° increments.
- Connection Type:Socket
- Port Size:20 mm socket
- Seal material:Viton®
- Description:PP Metering Ball Valve, 20 mm Socket
- Seq:3
- Environmentally Preferable:
- Wetted materials:Polypropylene
- Pk:1
- Inlet port:20 mm socket
- Outlet port:20 mm socket