Order Entry
Masterflex® Stepping-Motor Proportioning Valve, SS; 3/4"NPT(F), 1.5-10 VDC
  MFLX98651-10
undefined
Masterflex® Stepping-Motor Proportioning Valve, SS; 3/4"NPT(F), 1.5-10 VDC
  MFLX98651-10
1 080,00 €
  1 * 1 ST
  • Model:
  • 98651-10
  • 98651-08
  • 98651-04
  • 98651-02
  • 98651-00
  • 98651-06
  • Height:
  • 6,8"
  • 6,8"
  • 6,8"
  • 6,4"
  • 6,4"
  • 6,8"
  • Max. pressure:
  • 500 psi
  • 500 psi
  • 500 psi
  • 500 psi
  • 500 psi
  • 500 psi
  • Power:
  • 12 VDC
  • 12 VDC
  • 12 VDC
  • 12 VDC
  • 12 VDC
  • 12 VDC
  • Tubing ext.Ø:
  • 0,5"
  • 0,375"
  • 0,375"
  • 0,5"
  • Wetted materials:
  • SS, Viton, PFA
  • Aluminum, Viton, PFA
  • Aluminum, Viton, PFA
  • SS, Viton, PFA
  • Aluminum, Viton, PFA
  • SS, Viton, PFA
  • Width:
  • 3"
  • 3"
  • 5,4"
  • 5,2"
  • 5,2"
  • 5,4"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  •  
Resist pressures up to 500 psi!
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.
  • Connection Type:
    NPT(F)
  • Port Size:
    0,75 NPT(F)
  • Max. pressure:
    500 psi
  • Power:
    12 VDC
  • Description:
    Stepping-Motor Proportioning Valve, SS; 3/4"NPT(F), 1,5-10 VDC
  • Seq:
    06
  • Environmentally Preferable:
  • Model:
    98651-10
  • Wetted materials:
    SS, Viton, PFA
  • Width:
    3"
  • Height:
    6,8"
  • Warranty:
    1 year
  • Depth:
    50,8 mm
  • Pk:
    1
  • Cv:
    1,735
  • Inlet port:
    3/4" NPT(F)
  • Max. flow rate for air:
    1000 L/min
  • Max. flow rate for water:
    28 L/min
  • Outlet port:
    3/4" NPT(F)