Order Entry
Abaque HDN20 Replacement Hose, CSM, 0.79" ID x 1.46" OD; 29.72" L
  MFLX75810-36
CE Compliant
undefined
Abaque HDN20 Replacement Hose, CSM, 0.79" ID x 1.46" OD; 29.72" L
  MFLX75810-36
CE Compliant
646,00 €
  1 * 1 ST
  • Tubing int.Ø:
    20,00 mm (0,787")
  • Max. pressure:
    16 bar (235 psi)
  • Max. temperature:
    80 °C (175 °F)
  • Colour:
    Black
  • Description:
    CSM hose, green sticker, for use in HDN20 hose pumps
  • Length:
    0,755 m
  • Environmentally Preferable:
  • Length per pack:
    0,755 m (2,47 ft.)
  • Warranty:
    1 year
  • Working pressure:
    235 psi
  • Clarity:
    Opaque
  • Tubing material:
    CSM
  • Pk:
    1
  • Tubing int.Ø:
    20,00 mm (0,787")
  • Max. pressure:
    16 bar (235 psi)
  • Max. temperature:
    80 °C (175 °F)
  • Colour:
    Black
  • Description:
    CSM hose, green sticker, for use in HDN20 hose pumps
  • Length:
    0,755 m
  • Environmentally Preferable:
  • Length per pack:
    0,755 m (2,47 ft.)
  • Warranty:
    1 year
  • Working pressure:
    235 psi
  • Clarity:
    Opaque
  • Tubing material:
    CSM
  • Pk:
    1