6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one

Supplier: Thermo Scientific

Synonyms: 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

H62168.03EA 545 EUR
H62168.03 H62168.MD
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazin-3(4H)-one
Formula: C14H18BNO4
Storage Temperature: Ambient
MDL Number: MFCD12755796
CAS Number: 943994-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR