Food inspection case, EB 4401
Supplier: Ebro
620-1715EA 1660 EUR
620-1715
Food inspection case, EB 4401
Thermometers Digital Thermometers Digital Thermometers with Probe
This new standard food inspection case contains the required handhelds, data loggers and tools for comprehensive food inspections.
  • Frying oil quality measurement device FOM 320
  • Conformity valued thermometer TFX 422C
  • pH measurement device PHT 810 incl. accessories (penetration electrode, buffer solution)
  • Dual infrared /fold-back thermometer TLC 730
  • EBI 300 USB temperature data logger with Winlog.basic evaluation software
  • Flashlight
  • Knife, tweezers, scissors and magnifying glass
Plus About VWR