1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one

Supplier: Apollo Scientific
Danger

Synonyms: Allopurinol

APOSOR3110-25GEA 107 EUR
APOSOR3110-25G APOSOR3110-250G
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
Formula: C₅H₄N₄O
MW: 136.11
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00599413
CAS Number: 315-30-0
EINECS: 206-250-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR