Precision digital thermometers, testo 922 and 925
Supplier: Testo
620-1203EA 162 EUR
620-1203 620-1204
Precision digital thermometers, testo 922 and 925
Thermometers Digital Thermometers Precision Digital Thermometers
Digital thermometers with K-type connections.
  • 'HOLD' button to freeze reading
  • Continuous display of maximum/minimum values
  • Data can be sent directly to a printer
Model 922 (620-1203) Two-channel differential thermometer records temperature values from two connected thermocouple probes.
Model 925 (620-1204) One channel for fast action thermocouple probes has an audible alarm when limits are exceeded.
Delivery information: Supplied with nine volt battery and calibration protocol. Probe is not included - additional probes, including radio probes are avilable on request. A suitable printer also needs to be ordered separately. Please contact your local VWR office for details.
According to standard EN 60584-2, the accuracy of Class 1 refers to −40…+1000 °C (Type K), Class 2 to −40…+1200 °C (Type K), Class 3 to −200…+40 °C (Type K).
Plus About VWR