Freezers, storage appliances
Supplier: Liebherr
472-2251EA 0 EUR
LIEBUTSD3702 LIEBTGSD3702
Freezers, storage appliances
These freezer are optimal for the storage of frozen products at low temperatures, the glass-door display freezers comes with a facility of fan-assisted cooling for optimum presentation of goods. In conjunction with the interior light, the illuminated display is very eye-catching. The heated outer pane ensures that the insulation glass of the triple-glazed door does not mist up.
  • The glass doors gives an opportunity to organize the storage of frozen food in the most optimal way
  • In conjunction with the interior light, the illuminated display is very eye-catching
  • The heated outer pane ensures that the insulation glass of the triple-glazed door does not mist up
Temperature range: −10 to −22 °C