Imprimer
Bottles, wide neck, PTFE, with screw cap
Fournisseur: Vitlab
VITL122897EA 118 EUR
VITL122897 VITL122597 VITL122797 VITL122697
Bottles, wide neck, PTFE, with screw cap
PTFE, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Thick walled
With very wide mouth, ideal for filling with powders and paste-like materials.