5-Chloro-3-phenyl-2,1-benzisoxazole ≥98%
Supplier: Alfa Aesar
Warning
B23130.30EA 212 EUR
B23130.30 B23130.09 B23130.18
5-Chloro-3-phenyl-2,1-benzisoxazole ≥98%
5-Chloro-3-phenyl-2,1-benzisoxazole
Formula: C₁₃H₈ClNO
MW: 229.67
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00014573
CAS Number: 719-64-2
EINECS: 211-950-2