TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Synonyms: TBTU
ALEXALX121002G500EA 837 EUR
ALEXALX121002G500
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula: C₁₁H₁₆BF₄N₅O
MW: 321.1 g/mol
MDL Number: MFCD00077413
CAS Number: 125700-67-6