TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Supplier: Novabiochem (Merck biosciences)
|
Danger
|
Synonyms:
TBTU
01-62-0015-100EA
155
EUR
01-62-0015-100
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula:
C₁₁H₁₆BF₄N₅O MW: 321.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077413 CAS Number: 125700-67-6 EINECS: 423-040-4 UN: 1325 ADR: 4.1,II |