2-(3-Pyridinyl)-4-thiazolecarboxylic acid ≥97%
Supplier: Alfa Aesar
Warning
H51838.06EA 335 EUR
H51838.06 H51838.03
2-(3-Pyridinyl)-4-thiazolecarboxylic acid ≥97%
2-(3-Pyridinyl)-4-thiazolecarboxylic acid
Formula: C₉H₆N₂O₂S
MW: 206.22
Storage Temperature: Ambient
MDL Number: MFCD00052304
CAS Number: 39067-29-3