1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥99%
Danger
H27411.22EA 264 EUR
H27411.22
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥99%
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
Formula: C₁₉H₂₁ClN₂
Melting Pt: 261…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00035361
CAS Number: 71258-18-9
EINECS: 275-305-7