Methyl-2-pyrazinecarboxylate ≥98%
Supplier: Alfa Aesar
Warning
A12028.22EA 644 EUR
A12028.22 A12028.14 A12028.06
Methyl-2-pyrazinecarboxylate ≥98%
Methyl 2-pyrazinecarboxylate
Formula: C₆H₆N₂O₂
MW: 138.13
Melting Pt: 56…60 °C
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014611
CAS Number: 6164-79-0
EINECS: 228-198-6