1-Phenyl-1H-pyrazole-5-carbaldehyde

MAYBCC24504DAEA 0 EUR
MAYBCC24504DA MAYBCC24504CB MAYBCC24504DE
1-Phenyl-1H-pyrazole-5-carbaldehyde
1-Phenyl-1H-pyrazole-5-carbaldehyde
Formula: C₁₀H₈N₂O
MDL Number: MFCD00103497
CAS Number: 132274-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR