DL-Penicillamine

Supplier: TCI EUROPE

TCIAP0025-5GEA 0 EUR
TCIAP0025-5G TCIAP0025-25G
DL-Penicillamine
DL-Penicillamine
Formula: C₅H₁₁NO₂S
MW: 149.21
MDL Number: MFCD00004856
CAS Number: 52-66-4
EINECS: 200-147-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR