N-Chloroacetanilide ≥98.0% (by total nitrogen basis)
Supplier: TCI
|
Danger
|
Synonyms:
N-Chloroacetanilide
C2175-1G
TCIAC2175-1GEA
263
EUR
TCIAC2175-1G
N-Chloroacetanilide ≥98.0% (by total nitrogen basis)
NChloroacetanilide
Formula:
C₈H₈ClNO MW: 169.608 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01671408 CAS Number: 579-11-3 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...