Ethyl phenoxyacetate 98%
Supplier: Acros Organics
ACRO334941000EA 123 EUR
ACRO334941000 ACRO334940050
Ethyl phenoxyacetate 98%
Ethyl phenoxyacetate
Formula: C₁₀H₁₂O₃
MW: 180.20
Boiling Pt: 136 °C (19 mmHg)
Density: 1.1 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00026895
CAS Number: 2555-49-9
EINECS: 219-867-3
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5035 to 1.5065 (20°C, 589 nm)
GC ≥97.5 %