Imprimer

5-Chloro-N-(3-fluorobenzyl)-2-methylaniline, 97%

H57934.MDEA 33.1 EUR
ALFAH57934.MD ALFAH57934.03
5-Chloro-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1036621-04-1
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
MDL No.: MFCD11118340
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR