Imprimer

4-Ethyl-N-(3-fluorobenzyl)aniline, 97%

H57960.03EA 287 EUR
ALFAH57960.03 ALFAH57960.MD
4-Ethyl-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019492-41-1
Molecular Formula: C15H16FN
Formula Weight: 229.30
MDL No.: MFCD11146035
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR