2-Fluoro-N-(2-fluorobenzyl)-4-methylaniline, 97%

H57308.03EA 344 EUR
ALFAH57308.03 ALFAH57308.MD
2-Fluoro-N-(2-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1039823-53-4
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR