2-Fluoro-N-(4-fluorobenzyl)-4-methylaniline, 97%

H57078.03EA 246 EUR
ALFAH57078.03 ALFAH57078.MD
2-Fluoro-N-(4-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1040049-60-2
Molecular Formula: C14H13F2N
Formula Weight: 233.26
MDL No.: MFCD11181476
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR