4-Fluoro-N-(2-fluorobenzyl)-2-methylaniline, 97%

H57136.MDEA 71.7 EUR
ALFAH57136.MD ALFAH57136.03
4-Fluoro-N-(2-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1021080-27-2
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR