2,5-Difluoro-N-(3-fluorobenzyl)aniline, 97%
H57032.MDEA 71.7 EUR
ALFAH57032.MD ALFAH57032.03
2,5-Difluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019635-11-0
Molecular Formula: C13H10F3N
Formula Weight: 237.22
MDL No.: MFCD11142682