3-Fluoro-N-(4-fluorobenzyl)aniline ≥97%
Supplier: Thermo Scientific
Warning
H56989.03EA 553 EUR
H56989.03 H56989.MD
3-Fluoro-N-(4-fluorobenzyl)aniline ≥97%
3-Fluoro-N-(4-fluorobenzyl)aniline
Formula: C₁₃H₁₁F₂N
MW: 219.23
Storage Temperature: Ambient
MDL Number: MFCD11144475
CAS Number: 1019613-18-3