4-Fluoro-3-(trifluoromethoxy)benzonitrile ≥97%
Supplier: Thermo Scientific
Warning
H26667.03EA 131 EUR
H26667.03 H26667.06
4-Fluoro-3-(trifluoromethoxy)benzonitrile ≥97%
4-Fluoro-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD06660253
CAS Number: 886501-14-0
UN: 3276
ADR: 6.1,III