3-(3-Fluoro-5-(trifluoromethoxy)phenyl)propionic acid 97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-[3-Fluoro-5-(trifluoromethoxy)phenyl]propanoic acidBenzenepropanoic acid, 3-fluoro-5-(trifluoromethoxy)-
H34223.MDEA 50.7 EUR
H34223.MD H34223.03
3-(3-Fluoro-5-(trifluoromethoxy)phenyl)propionic acid 97%
3-(3-Fluoro-5-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₈F₄O₃
MW: 252.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783232
CAS Number: 1261828-37-8