4,5-Difluoro-o-toluidine ≥99%
Danger
Synonyms: 4,5-Difluoro-2-methylaniline
H26142.MDEA 57.2 EUR
H26142.MD H26142.03
4,5-Difluoro-o-toluidine ≥99%
4,5-Difluoro-o-toluidine
Formula: C₇H₇F₂N
MW: 143.14
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08062378
CAS Number: 875664-57-6
EINECS: None
UN: 2811
ADR: 6.1,III