3-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%
Supplier: Alfa Aesar
Warning
H32430.03EA 62.3 EUR
H32430.03 H32430.06
3-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Methoxy-5-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD09025409
CAS Number: 53985-48-1