4-(2-Chloroethyl)anisole 97%
|
Warning
|
Synonyms:
1-(2-Chloroethyl)-4-methoxybenzene
ACRO123110050EA
22.2
EUR
ACRO123110050
4-(2-Chloroethyl)anisole 97%
4-(2-Chloroethyl)anisole
Formula:
C₉H₁₁ClO Boiling Pt: 125…128 °C (13mbar) Density: 1.13 g/cm³ Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044718 CAS Number: 18217-00-0 EINECS: 242-099-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5360 to 1.5380 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific gravity | (20°C) 1.12 to 1.13 |