(±)-Aspartic acid
Supplier: Molekula
Synonyms: DL-Aspartic acid2-Aminobutanedioic acid
MOLE22164347-1KGEA 0 EUR
MOLE22164347-1KG
(±)-Aspartic acid
(±)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 210-513-3