Bis(2-methoxyethyl)amine ≥98.0% (by GC, titration analysis)
Supplier: TCI EUROPE
|
Danger
|
TCIAB1383-25MLEA
66
EUR
TCIAB1383-25ML
Bis(2-methoxyethyl)amine ≥98.0% (by GC, titration analysis)
Bis(2-methoxyethyl)amine
Formula:
C₆H₁₅NO₂ Boiling Pt: 172°C Flash Pt: 58°C |
MDL Number:
MFCD00025906 CAS Number: 111-95-5 EINECS: 203-923-9 UN: 2734 ADR: 8,II |
![]() |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |