(S)-(+)-1-Benzyl-3-hydroxypiperidine ≥97%
Supplier: Alfa Aesar
Warning
H30615.MDEA 60.5 EUR
H30615.MD H30615.03
(S)-(+)-1-Benzyl-3-hydroxypiperidine ≥97%
(S)-(+)-1-Benzyl-3-hydroxypiperidine
Formula: C₁₂H₁₇NO
MDL Number: MFCD03426359
CAS Number: 91599-79-0