1-Bromo-2-butene ≥83.0% (by GC)
Supplier: TCI EUROPE
|
Danger
|
Synonyms:
1-Bromo-2-butene
, Crotyl bromide
TCIAB0562-25MLEA
184
EUR
TCIAB0562-25ML
1-Bromo-2-butene ≥83.0% (by GC)
1-Bromo-2-butene
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 107°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000248 CAS Number: 4784-77-4 EINECS: 225-332-5 UN: 1993 ADR: 3,II |
![]() |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 83.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...