3-Methyl-1-pentyne-3-ol ≥98.0% (by GC)

Supplier: TCI EUROPE
Warning

Synonyms: 3-Hydroxy-3-methyl-1-pentyneEthinyl ethyl methyl carbinol3-Methylpent-1-yn-3-ol3-Methyl-1-pentyn-3-ol

TCIAM0396-25MLEA 17.6 EUR
TCIAM0396-25ML
3-Methyl-1-pentyne-3-ol ≥98.0% (by GC)
3-Methyl-1-pentyne-3-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 122°C
Melting Pt: –30 °C
Flash Pt: 38°C
MDL Number: MFCD00004479
CAS Number: 77-75-8
EINECS: 201-055-5
UN: 1987
ADR: 3,III
Merck Index: 13,05870
https://be.vwr.com/stibo/web/std.lang.all/38/18/28973818.jpg

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR