Benzyl salicylate

Supplier: Roth Carl

Synonyms: Benzyl 2-hydroxybenzoate

ROTH4184.1EA 42.9 EUR
ROTH4184.1
Benzyl salicylate
Benzyl salicylate
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 23.9 °C
Density: 1.1755 g/cm³ (20 °C)
MDL Number: MFCD00020034
CAS Number: 118-58-1
EINECS: 204-262-9
Merck Index: 13,01146

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR