2-Iodo-2-methylpropane ≥97.0% (by GC) stabilised

Supplier: TCI EUROPE
Danger

Synonyms: tert-Butyl iodide2-Methyl-2-propyliodide

Stabilization: Stabilised with copper chip

TCIAB1072-25GEA 61.6 EUR
TCIAB1072-25G
2-Iodo-2-methylpropane ≥97.0% (by GC) stabilised
2-Iodo-2-methylpropane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 99°C
Melting Pt: –78 °C
Flash Pt: -10°C
Storage Temperature: Refrigerator
MDL Number: MFCD00001068
CAS Number: 558-17-8
EINECS: 209-190-1
UN: 2391
ADR: 3,II
https://be.vwr.com/stibo/web/std.lang.all/29/28/28932928.jpg

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR