2,5-Dichloroisonicotinic acid 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2,5-Dichloropyridine-4-carboxylic acid
H58277.03EA 61.7 EUR
H58277.03 H58277.MD
2,5-Dichloroisonicotinic acid 95%
2,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 235…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00466640
CAS Number: 88912-26-9