Methylcyclopentadiene dimer (so called) stabilised

Supplier: TCI EUROPE
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

TCIAM0920-25MLEA 17.6 EUR
TCIAM0920-25ML
Methylcyclopentadiene dimer (so called) stabilised
Methylcyclopentadiene dimer
Formula: C₁₂H₁₆
MW: 160.26 g/mol
Flash Pt: 53°C
MDL Number: MFCD00082297
CAS Number: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III
https://be.vwr.com/stibo/web/std.lang.all/44/67/28974467.jpg

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR