(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)
Supplier: TCI EUROPE
|
Warning
|
Synonyms:
2,2-Diethoxy-N,N-dimethylethanamine
TCIAD1703-25MLEA
80.3
EUR
TCIAD1703-25ML
(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)
(Dimethylamino)acetaldehyde diethylacetal
Formula:
C₈H₁₉NO₂ MW: 161.25 g/mol Boiling Pt: 60°C Flash Pt: 45°C |
MDL Number:
MFCD00009232 CAS Number: 3616-56-6 EINECS: 222-800-0 UN: 1993 ADR: 3,III |
![]() |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...