4-Fluoro-3-iodobenzaldehyde ≥97%
Warning
H33615.MDEA 129 EUR
H33615.MD H33615.03
4-Fluoro-3-iodobenzaldehyde ≥97%
4-Fluoro-3-iodobenzaldehyde
Formula: C₇H₄OFI
Melting Pt: 65…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08236850
CAS Number: 227609-88-3