Imprimer
Safety shoes, lace-up, Jalarthur
Fournisseur: Jallatte
JALCJMI04/47EA 92.3 EUR
JALCJMI04/47 JALCJMI04/46 JALCJMI04/45 JALCJMI04/44 JALCJMI04/43 JALCJMI04/42 JALCJMI04/41 JALCJMI04/40 JALCJMI04/39
Safety shoes, lace-up, Jalarthur
Metal-free safety shoes made from full grain leather upper, leather lining and Softane™ dual-density PU sole (resistant to abrasion, hydrocarbons, oils and grease).
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
Colour: Black
Certifications: EN ISO 20345:2004; S1P, SRC