Thermal mixers, ThermoMixer® F1.5 and FP
Supplier: Eppendorf
Thermomixer®
444-1024EA 2533 EUR
521-2112 444-1024 444-1023 EPPE5385000.024 460-0225 460-0224 EPPE5384000.020
Thermal mixers, ThermoMixer® F1.5 and FP
Shakers Shaking Incubators
Eppendorf ThermoMixer® F1.5 is customised for 1,5 ml reaction vessels and Eppendorf ThermoMixer® FP is designed for plates. Units feature simple and intuitive operation using predefined temperature keys (37, 42, 56 and 95 °C). Both units are ideal for bacterial growth, enzymatic reactions and lysis reactions at  100 °C. The F1.5 is suitable for denaturation of DNA, RNA and proteins, labelling of nucleic acids and proteins and Proteinase K digestions of cells and tissues. The FP model is suitable for resuspending pellets, mixing protein quantification assays and ELISA assays.
  • Precise temperature control from ambient +4 to 100 °C
  • Efficient mixing up to 1500 min⁻¹ ( F1.5) or 2000 min⁻¹ (FP) thanks to 2DMix-Control and anti-spill technology
  • Digital display enables simple and accurate setting and viewing of parameters
  • Prevent condensation on tube lids and walls and improve temperature homogeneity with the new ThermoTop®
Ordering information: Eppendorf ThermoMixer® F1.5 is supplied with fixed SmartBlock™ for 24×1,5 ml reaction vessels, and Eppendorf ThermoMixer® FP is supplied with fixed SmartBlock™ for microplates and deepwell plates, including lid. Can be used with ThermoTop® cover.