4-Chloro-N-cyclohexylbenzamide ≥97%
|
Warning
|
H55146.03EA
478
EUR
H55146.03
4-Chloro-N-cyclohexylbenzamide ≥97%
4-Chloro-N-cyclohexylbenzamide
Formula:
C₁₃H₁₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00029412 CAS Number: 57707-20-7 |